missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium molybdenum oxide, anhydrous, 99.8% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Potassium molybdenum oxide used in chemical research, Chemicals, pharmaceutical research, pharmaceutical intermediates reagents, Laboratory chemicals, Manufacture of substances. And it is an important raw material and intermediates used in organic synthesis, agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 919°C |
Color | White |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.8% (metals basis) |
Solubility Information | Soluble in water. |
IUPAC Name | dipotassium dioxomolybdenumbis(olate) |
Formula Weight | 238.14 |
Odor | Odorless |
Appearance | White powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'