Learn More
Potassium molybdenum oxide, anhydrous, 99.8% (metals basis), Thermo Scientific Chemicals
£44.42 - £160.80
Chemical Identifiers
CAS | 13446-49-6 |
---|---|
Molecular Formula | K2MoO4 |
Molecular Weight (g/mol) | 238.14 |
MDL Number | MFCD00011370 |
InChI Key | NUYGZETUIUXJCO-UHFFFAOYSA-N |
Synonym | potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous |
PubChem CID | 6093816 |
SMILES | [K+].[K+].[O-][Mo]([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11328127
|
Thermo Scientific Alfa Aesar
022898.09 |
10 g |
£44.42
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338127
|
Thermo Scientific Alfa Aesar
022898.18 |
50 g |
£160.80
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium molybdenum oxide used in chemical research, Chemicals, pharmaceutical research, pharmaceutical intermediates reagents, Laboratory chemicals, Manufacture of substances. And it is an important raw material and intermediates used in organic synthesis, agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13446-49-6 | |
238.14 | |
NUYGZETUIUXJCO-UHFFFAOYSA-N | |
6093816 |
K2MoO4 | |
MFCD00011370 | |
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous | |
[K+].[K+].[O-][Mo]([O-])(=O)=O |
Specifications
919°C | |
White | |
99.8% (metals basis) | |
MFCD00011370 | |
Soluble in water. | |
[K+].[K+].[O-][Mo]([O-])(=O)=O | |
238.14 | |
238.14 | |
White powder | |
Potassium molybdenum oxide, anhydrous |
13446-49-6 | |
Powder | |
K2MoO4 | |
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous | |
NUYGZETUIUXJCO-UHFFFAOYSA-N | |
dipotassium dioxomolybdenumbis(olate) | |
6093816 | |
Odorless | |
2.3 |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 236-599-2
missing translation for 'rtecsNumber' : QA4930000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only