missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
250 g
5 kg
Description
KNO3, CAS Number-7757-79-1, nitre, saltpeter, nitric acid potassium salt, kaliumnitrat, nitrate of potash, vicknite, nitric acid, potassium salt, kalii nitras, potassium nitrate, saltpetre, 500g, 400 deg.C, CHEBI:63043, White, 101.1g/mol, FGIUAXJPYTZDNR-UHFFFAOYSA-N, potassium;nitrate, 334 deg.C
Specifications
Boiling Point | 400°C |
Melting Point | 334°C |
Color | White |
pH | 6 to 8 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 101.1g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium nitrate
- Warning
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'