missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrate, Extra Pure, SLR, Fisher Chemical™
£420.00
Chemical Identifiers
CAS | 7757-79-1 |
---|---|
Molecular Formula | KNO3 |
Molecular Weight (g/mol) | 101.10 |
MDL Number | MFCD00011409 |
InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras |
PubChem CID | 24434 |
ChEBI | CHEBI:63043 |
SMILES | [K+].[O-][N+]([O-])=O |
Description
Chemical Identifiers
7757-79-1 | |
101.10 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
24434 | |
[K+].[O-][N+]([O-])=O |
KNO3 | |
MFCD00011409 | |
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
CHEBI:63043 |
Specifications
400°C | |
7757-79-1 | |
6 to 8 | |
KNO3 | |
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
[K+].[O-][N+]([O-])=O | |
101.10 | |
CHEBI:63043 | |
Plastic powder jar |
334°C | |
White | |
Solid | |
MFCD00011409 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
potassium nitrate | |
24434 | |
101.1g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium nitrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.