missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrate, Puratronic™, 99.995% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
5g
50g
250g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 5g |
Assay Percent Range | 99.998% (metals basis) |
Solubility Information | Moderately soluble in water,and insoluble in alcohol. Soluble in glycerol and ammonia. |
Formula Weight | 101.11 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White crystalline powder |
Vapor Pressure | 1.33 hpa (1mm Hg) at 745°C |
Density | 2.109 |
Chemical Name or Material | Potassium nitrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'