missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 kg |
UN Number | 1486 |
IUPAC Name | potassium;nitrate |
Formula Weight | 101.1g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Potassium Nitrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
Product Identifier
- Potassium nitrate
- Warning
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'