missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate, Technical, Fisher Chemical™

£48.80 - £138.00

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10598440
View Documents Promotion Details
Fisher Chemical
P/6040/60
1 kg
£48.80
1kg
Please to purchase this item. Need a web account? Register with us today!
10578630
View Documents Promotion Details
Fisher Chemical
P/6040/63
3 kg
£138.00
3kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
[K+].[O-][N+]([O-])=O
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043

Specifications

400°C
7757-79-1
6 to 8
KNO3
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
[K+].[O-][N+]([O-])=O
101.10
CHEBI:63043
Plastic powder jar
334°C
White
Solid
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
24434
101.1g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium nitrate

Signal Word
  • Warning

Hazard Category
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.

SDS
Documents

Documents

Certificates