missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrite, 97%, extra pure, Thermo Scientific Chemicals
£60.60 - £272.36
Chemical Identifiers
CAS | 7758-09-0 |
---|---|
Molecular Formula | KNO2 |
Molecular Weight (g/mol) | 85.10 |
MDL Number | MFCD00011408 |
InChI Key | BXNHTSHTPBPRFX-UHFFFAOYSA-M |
Synonym | potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 |
PubChem CID | 516910 |
SMILES | [K+].[O-]N=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15113908
|
Thermo Scientific Acros
453190010 |
1 kg |
£60.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15289315
|
Thermo Scientific Acros
453190050 |
5 kg |
£272.36
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-09-0 | |
85.10 | |
BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
516910 |
KNO2 | |
MFCD00011408 | |
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 | |
[K+].[O-]N=O |
Specifications
387.0°C | |
97% | |
KNO2 | |
15,777 | |
Solubility in water: 3000g/L water (20°C). Other solubilities: 413 g/100mL water (100°C), slightly soluble in alcohol, soluble in hot alcohol, very soluble in liquid nh3 | |
[K+].[O-]N=O | |
85.10 | |
85.1 | |
Extra Pure |
7758-09-0 | |
KNO2 | |
MFCD00011408 | |
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 | |
BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
potassium nitrite | |
516910 | |
97% | |
Potassium nitrite, Extra Pure |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-832-4
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium nitrite
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
- MIXTURE LIST-Contains : Potassium nitrite
RUO – Research Use Only