missing translation for 'onlineSavingsMsg'
Learn More

Potassium orthovanadate, 99.9% (metals basis)

Product Code. 11318308
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11318308

Brand: Thermo Scientific Alfa Aesar 039502.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14293-78-8 | K3O4V | 232.232 g/mol

It is employed as in chemical fertilizer industries and used as corrosion inhibitor. It is also used for ammonia synthesis. Used as a catalyst in oil & gas industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14293-78-8
Molecular Formula K3O4V
Molecular Weight (g/mol) 232.232
MDL Number MFCD00210642
InChI Key FBRINGOENSVSPU-UHFFFAOYSA-N
Synonym Potassium vanadate (ortho)
PubChem CID 22023817
IUPAC Name tripotassium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Physical Form Granules
Quantity 100 g
UN Number UN3285
Solubility Information Soluble in water.
Formula Weight 232.24
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium orthovanadate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.