missing translation for 'onlineSavingsMsg'
Learn More

Potassium osmium(VI) oxide dihydrate, 98+%, Thermo Scientific Chemicals

£68.14 - £194.97

Chemical Identifiers

CAS 10022-66-9
Molecular Formula H4K2O6Os
Molecular Weight (g/mol) 368.453
MDL Number MFCD00149919
InChI Key DGODWNOPHMXOTR-UHFFFAOYSA-N
Synonym potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate
PubChem CID 53393272
IUPAC Name dipotassium;dioxido(dioxo)osmium;dihydrate
SMILES O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377487
View Documents
Thermo Scientific Alfa Aesar
012647.01
0.25 g
£68.14
0.25g
Please to purchase this item. Need a web account? Register with us today!
11367487
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012647.03
1 g
£194.97
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium osmium(VI) oxide dihydrate acts as a catalyst for the oxidative cleavage of olefins to ketones and carboxylic acids. It is employed in an intramolecular amidohydroxylation of carbamoyloxytethered olefins providing hydroxy oxazolidinones. It is also used as a reagent in the synthesis of amphidinolide B, which shows potent antitumor activity against human solid and blood tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10022-66-9
368.453
DGODWNOPHMXOTR-UHFFFAOYSA-N
53393272
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]
H4K2O6Os
MFCD00149919
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate
dipotassium;dioxido(dioxo)osmium;dihydrate

Specifications

10022-66-9
Powder
H4K2O6Os
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate
DGODWNOPHMXOTR-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)osmium;dihydrate
53393272
Hygroscopic
Potassium osmium(VI) oxide dihydrate
White
99+%
MFCD00149919
Soluble in water. Slowly decomposes in aqueous solution forming the tetroxide. Insoluble in alcohol,etherSoluble in water. Insoluble in alcohol and ether.
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]
368.453
368.43 (332.40 Anhydrous)
Purple powder
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c

H272-H290-H312+H332-H314-H335

missing translation for 'dotInformation' : DOT Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 243-247-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P405-Store locked up.

SDS
Documents

Documents

Certificates

RUO – Research Use Only