missing translation for 'onlineSavingsMsg'
Learn More
Potassium oxalate monohydrate, 99%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10209420

Potassium oxalate monohydrate, 99%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10209420
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10209420

Brand: Thermo Scientific Acros 424025000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium;oxalate;hydrate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 356.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula KO2CCO2K·H2O
Quantity 500 g
Merck Index 15, 7772
Solubility Information Solubility in water: 364g/L (20°C)
Formula Weight 184.24
Percent Purity 99%
Grade ACS Reagent
Insoluble Matter 0.01% max
Physical Form Crystalline Powder
Chemical Name or Material Potassium oxalate monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464546 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog Number42402016,424025000,424020025
Lot NumberA0399784 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number42402016,424025000,424020025
Lot NumberA0418440 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number42402016,424025000,424020025
Lot NumberA0446029 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog Number42402016,424025000,424020025
Lot NumberA0445532 Certificate TypeCertificate of Analysis Date31/08/2022 Catalog Number42402016,424025000,424020025
compliance-icons
Product Identifier
  • Potassium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium oxalate monohydrate, 99%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.