missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate Monohydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10000310
Click to view available options
Quantity:
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 356°C
Color White
pH 7.0 to 8.5
Physical Form Solid
Quantity 500 g
IUPAC Name dipotassium hydrate oxalate
Formula Weight 184.24g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Potassium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'