missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Perchlorate, ≥98.5% (T), purum p.a, Honeywell™
Purum p.a.,≥98.5% (T)
£160.53 - £380.51
Chemical Identifiers
CAS | 7778-74-7 |
---|---|
Molecular Formula | ClKO4 |
Molecular Weight (g/mol) | 138.54 |
MDL Number | MFCD00011362 |
InChI Key | YLMGFJXSLBMXHK-UHFFFAOYSA-M |
Synonym | potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp |
PubChem CID | 516900 |
IUPAC Name | potassium;perchlorate |
SMILES | [K+].[O-][Cl](=O)(=O)=O |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
7778-74-7 | |
138.54 | |
YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
516900 | |
[K+].[O-][Cl](=O)(=O)=O |
ClKO4 | |
MFCD00011362 | |
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
potassium;perchlorate |
Specifications
>400°C | |
White | |
ClKO4 | |
MFCD00011362 | |
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
[K+].[O-][Cl](=O)(=O)=O | |
138.54 | |
138.55g/mol | |
Odorless | |
Potassium perchlorate |
7778-74-7 | |
Crystalline | |
KClO4 | |
UN1489 | |
YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
potassium;perchlorate | |
516900 | |
≥98.5% (T) | |
2.52g/cm3 at 20°C |
Safety and Handling
P220
missing translation for 'dotInformation' : 5.1 / PGII
missing translation for 'rtecsNumber' : 231-912-9