missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium perchlorate, ACS reagent, Thermo Scientific Chemicals
£22.61 - £64.22
Chemical Identifiers
CAS | 7778-74-7 |
---|---|
Molecular Formula | ClKO4 |
Molecular Weight (g/mol) | 138.54 |
MDL Number | MFCD00011362 |
InChI Key | YLMGFJXSLBMXHK-UHFFFAOYSA-M |
Synonym | potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp |
PubChem CID | 516900 |
IUPAC Name | potassium;perchlorate |
SMILES | [K+].[O-][Cl](=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12624967
|
Thermo Scientific Acros
221371000 |
100 g | Plastic bottle |
£22.61
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12634967
|
Thermo Scientific Acros
221375000 |
500 g | Plastic bottle |
£64.22
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-74-7 | |
138.54 | |
YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
516900 | |
[K+].[O-][Cl](=O)(=O)=O |
ClKO4 | |
MFCD00011362 | |
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
potassium;perchlorate |
Specifications
400.0°C | |
ClKO4 | |
MFCD00011362 | |
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
potassium;perchlorate | |
516900 | |
ACS Reagent |
7778-74-7 | |
KClO4 | |
15,7774 | |
Solubility in water: 17g/L (20°C). Other solubilities: 0.75 g/100mL water (0°C), 21.8 g/100mL water (100°C), practically insoluble in alcohol | |
[K+].[O-][Cl](=O)(=O)=O | |
138.54 | |
138.55 | |
Potassium perchlorate |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-912-9
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium perchlorate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Oxidising solid Category 1
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Potassium perchlorate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title