Learn More
Potassium periodate, 99.5%, extra pure, Thermo Scientific Chemicals
£37.39 - £533.66
Chemical Identifiers
CAS | 7790-21-8 |
---|---|
Molecular Formula | IKO4 |
Molecular Weight (g/mol) | 230 |
InChI Key | FJVZDOGVDJCCCR-UHFFFAOYSA-M |
Synonym | potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1 |
PubChem CID | 516896 |
IUPAC Name | potassium;periodate |
SMILES | [O-]I(=O)(=O)=O.[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10022510
|
Thermo Scientific Acros
198391000 |
100 g | Plastic bottle |
£37.39
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10296870
|
Thermo Scientific Acros
198395000 |
500 g | Plastic bottle |
£142.71
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11944221
|
Thermo Scientific Acros
198390025 |
2.5 kg | Plastic bottle |
£533.66
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7790-21-8 | |
230 | |
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1 | |
potassium;periodate |
IKO4 | |
FJVZDOGVDJCCCR-UHFFFAOYSA-M | |
516896 | |
[O-]I(=O)(=O)=O.[K+] |
Specifications
582.0°C | |
White | |
99.4% min. (Iodometry) | |
KIO4 | |
15,7775 | |
Solubility in water: 0.42 g/100mL (20°C). | |
[O-]I(=O)(=O)=O.[K+] | |
230 | |
230 | |
Extra Pure |
7790-21-8 | |
Crystalline Powder | |
IKO4 | |
01,809; 02,311; 04,373; 05,507; 06,455 | |
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1 | |
FJVZDOGVDJCCCR-UHFFFAOYSA-M | |
potassium;periodate | |
516896 | |
99.5% | |
Potassium periodate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May intensify fire; oxidizer.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occu
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-196-
- Potassium periodate
Signal Word
- Danger
Hazard Category
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 1
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity Category 1
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only