missing translation for 'onlineSavingsMsg'
Learn More

Potassium periodate, 99%, Thermo Scientific Chemicals

Product Code. 11493694
Click to view available options
Quantity:
100 g
500 g
2500 g

Potassium periodate is used as an oxidizing agent in organic synthesis and an analytical reagent for the determination of potassium and cerium. It is also used in colorimetric estimation of manganese. It reacts with ammonium cerium(IV) nitrate to prepare tris[trinitratocerium(IV)] paraperiodate which finds application as an oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium periodate is used as an oxidizing agent in organic synthesis and an analytical reagent for the determination of potassium and cerium. It is also used in colorimetric estimation of manganese. It reacts with ammonium cerium(IV) nitrate to prepare tris[trinitratocerium(IV)] paraperiodate which finds application as an oxidant.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong reducing agents, powdered metals and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 229.999
MDL Number MFCD00011407
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
PubChem CID 516896
IUPAC Name potassium;periodate
SMILES [O-]I(=O)(=O)=O.[K+]

Specifications

Melting Point 582°C
Quantity 100 g
UN Number UN1479
Merck Index 14,7654
Solubility Information Soluble in water.
Formula Weight 230
Percent Purity 99%
Density 3.618
Chemical Name or Material Potassium periodate
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'