missing translation for 'onlineSavingsMsg'
Learn More

Potassium periodate, ACS, 99.8%-100.3%

Product Code. p-4407757
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11388117

Brand: Thermo Scientific Alfa Aesar 022896.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-21-8 | IKO4 | 229.999 g/mol

For the colorimetric determination of Mn (oxidizes manganese compounds to permanganates), as oxidizing agentPotassium periodate is used as an oxidizing agent in organic synthesis and an analytical reagent for the determination of potassium and cerium. It is also used in colorimetric estimation of manganese. It reacts with ammonium cerium(IV) nitrate to prepare tris[trinitratocerium(IV)] paraperiodate which finds application as an oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 229.999
MDL Number MFCD00011407
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
PubChem CID 516896
IUPAC Name potassium;periodate
SMILES [O-]I(=O)(=O)=O.[K+]

Specifications

Melting Point 582°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.8%-100.3%
Solubility Information Soluble in water.
Formula Weight 230
Grade ACS Reagent
Density 3.618
Chemical Name or Material Potassium periodate, For ACS analysis
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.