missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Periodate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | KIO4 |
UN Number | 1479 |
Formula Weight | 230g/mol |
Percent Purity | 99% |
Grade | ACS Reagent |
Chemical Name or Material | Potassium Periodate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'