missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥99.0%
Chemical Identifiers
CAS | 7722-64-7 |
---|---|
Molecular Formula | KMnO4 |
Molecular Weight (g/mol) | 158.032 |
MDL Number | MFCD00011364 |
InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san sooShow More |
PubChem CID | 516875 |
IUPAC Name | potassium;permanganate |
SMILES | [O-][Mn](=O)(=O)=O.[K+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7722-64-7 | |
158.032 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
516875 | |
[O-][Mn](=O)(=O)=O.[K+] |
KMnO4 | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san sooShow More | |
potassium;permanganate |
Specifications
7722-64-7 | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
[O-][Mn](=O)(=O)=O.[K+] | |
158.032 | |
158.03g/mol | |
ACS Reagent |
KMnO4 | |
1490 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
potassium;permanganate | |
516875 | |
≥99% | |
Potassium Permanganate |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
H272-H302-H314-H410
missing translation for 'einecsNumber' : 231-760-3
missing translation for 'rtecsNumber' : SD6475000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium permanganate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : potassium permanganate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Permanganate, ACS Reagent, Honeywell Fluka™