missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Amber glass bottle
Glass Jar
Unit Size:
500g
5kg
Description
KMnO4, CAS Number-7722-64-7, solo san soo, chameleon mineral, cairox, walko tablets, argucide, potassium permanganate, insta-perm, algae-k, condy's crystals, permanganate of potash, 500g, Brown, 158.04g/mol, VZJVWSHVAAUDKD-UHFFFAOYSA-N, potassium;permanganate, 240 deg.C, 11364, 158.032, Glass Jar
Specifications
Specifications
| Melting Point | 240°C |
| Color | Brown |
| pH | 8 |
| Physical Form | Powder/Solid |
| Quantity | 5 kg |
| Formula Weight | 158.04g/mol |
| Packaging | Amber glass bottle |
Safety and Handling
Product Identifier
- Potassium permanganate
- Danger
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction