missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate, puriss p.a, ACS, ≥99.0% (RT), Honeywell Fluka™

Product Code. 15625980
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent, Hg ≤0.000005%,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Violet
Physical Form Crystalline
Quantity 1 kg
UN Number UN1490
Formula Weight 158.03g/mol
Percent Purity ≥99.0% (RT)
Odor Odorless
Packaging Glass bottle
Decomposition Information >240°C
Density 2.7g/cm3
Chemical Name or Material Potassium permanganate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'