missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate, puriss p.a, ACS, ≥99.0% (RT), Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | Violet |
Physical Form | Crystalline |
Quantity | 250 g |
UN Number | UN1490 |
Formula Weight | 158.03g/mol |
Percent Purity | ≥99.0% (RT) |
Odor | Odorless |
Packaging | Glass bottle |
Decomposition Information | >240°C |
Density | 2.7g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'