missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate Solution 0.02M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

£54.30 - £95.20

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number 11364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantity Packaging Prix Quantité et disponibilité  
Code produit Marque Quantity Packaging Prix Quantité et disponibilité  
10705721
missing translation for 'viewDocuments' Détails de la promotion
Fisher Chemical
J/6900/15
1 L Amber glass bottle
£54.30
1 L
Please to purchase this item. Need a web account? Register with us today!
10326190
missing translation for 'viewDocuments' Détails de la promotion
Fisher Chemical
J/6900/PB17
2.5 L Plastic Coated Glass bottle
£95.20
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium Permanganate Solution 0.02M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis

Spécification

Identifiants chimiques

7722-64-7
158.032
VZJVWSHVAAUDKD-UHFFFAOYSA-N
516875
[O-][Mn](=O)(=O)=O.[K+]
KMnO4
11364
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
potassium;permanganate

Spécification

7722-64-7
Liquid
11364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium;permanganate
516875
Purple
KMnO4
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
[O-][Mn](=O)(=O)=O.[K+]
158.032
Vidéos
Sécurité et traitement

Sécurité et traitement

Identifiant produit
  • potassium permanganate 0.02M

Mention d'avertissement
  • Warning

Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 3

Mention de danger
  • H412-Harmful to aquatic life with long lasting effects.

Conseils de prudence
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Informations supplémentaires
  • MIXTURE LIST-Contains : potassium permanganate

FDS
Documents

Documents

Certificats