missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate solution, Volumetric, 0.02 M KMnO4 (0.1N), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations

Specifications
Specifications
CAS | 7722-64-7 |
Quantity | 1 L |
Molecular Formula | KMnO4 |
MDL Number | MFCD00011364 |
UN Number | UN3082 |
Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
SMILES | [O-][Mn](=O)(=O)=O.[K+] |
IUPAC Name | potassium;permanganate |
Molecular Weight (g/mol) | 158.032 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling

Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : potassium permanganate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium permanganate solution, Volumetric, 0.02 M KMnO4 (0.1N), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction