missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium perrhenate, 99% (metals basis), Re 64%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.2 g
2 g
10 g
50 g
Description
A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1360°C to 1370°C |
Melting Point | 550°C |
Color | White |
Physical Form | Crystalline |
Quantity | 0.2 g |
UN Number | UN1479 |
Solubility Information | Soluble in water. |
Formula Weight | 289.3 |
Percent Purity | 99% |
Concentration or Composition (by Analyte or Components) | Re 64% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'