missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium perruthenate(VII), 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Potassium perruthenate(VII) is used in the production of carbon nanotubes for flexible solid-state supercapacitors. It is also used as an oxidizing agent in the synthesis of 5-formylcytosine from 5-hydroxymethylcytosine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 440°C (decomposition) |
Color | Black |
Physical Form | Crystalline |
Quantity | 1 g |
Assay Percent Range | 97% |
Solubility Information | Slightly soluble in water. |
Formula Weight | 204.17 |
Appearance | Black crystals |
Chemical Name or Material | Potassium perruthenate(VII) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'