missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Dibasic, ≥98%, ACS reagent, Honeywell Fluka™
ACS Reagent,≥98%
£21.43 - £3755.50
Chemical Identifiers
CAS | 7758-11-4 |
---|---|
Molecular Formula | HK2O4P |
Molecular Weight (g/mol) | 174.17 |
MDL Number | MFCD00011383 |
InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
Synonym | dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate |
PubChem CID | 24450 |
ChEBI | CHEBI:32031 |
IUPAC Name | dipotassium;hydrogen phosphate |
SMILES | OP(=O)([O-])[O-].[K+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15608020
|
Honeywell-Fluka
P3786-100G |
100 g |
£21.43
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15648020
|
Honeywell-Fluka
P3786-500G |
500 g |
£81.12
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15618020
|
Honeywell-Fluka
P3786-1KG |
1 kg |
£95.24
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15628020
|
Honeywell-Fluka
P3786-25KG |
2.5 kg |
£3755.50
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15658020
|
Honeywell-Fluka
P3786-6X500G |
6 x 500 g |
£550.56
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15638020
|
Honeywell-Fluka
P3786-4X2.5KG |
4 x 2.5 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7758-11-4 | |
174.17 | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
24450 | |
dipotassium;hydrogen phosphate |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
CHEBI:32031 | |
OP(=O)([O-])[O-].[K+].[K+] |
Specifications
7758-11-4 | |
K2HPO4 | |
NONH for all modes of transport | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate | |
24450 | |
174.18g/mol | |
Potassium phosphate dibasic |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
OP(=O)([O-])[O-].[K+].[K+] | |
174.17 | |
CHEBI:32031 | |
≥98% |