missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, dibasic, 98+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10331211

Potassium phosphate, dibasic, 98+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10331211
500 g, Plastic bottle
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10331211

Brand: Thermo Scientific Acros 424195000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.18
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphateShow MoreShow Less
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340.0°C
Color White
pH 8.5 to 9.6 (5% soln. at 25°C)
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 98+%
Linear Formula K2HPO4
Merck Index 15,7779
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 174.18
Percent Purity ≥98%
Grade ACS Reagent
Loss on Drying 1.0% max. (105°C)
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, dibasic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463738 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number424190100,424190050,424195000,424190025
Lot NumberA0435691 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number424190100,424190050,424195000,424190025
Lot NumberA0406912 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number424190100,424190050,424195000,424190025
Lot NumberA0431705 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number424190100,424190050,424195000,424190025
Lot NumberA0439663 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number424190100,424190050,424195000,424190025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, dibasic, 98+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.