missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals Product Code.: 11944291

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Product Code. 11944291
5 kg, Plastic drum
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 11944291

Brand: Thermo Scientific Acros 215470050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, ACROS Organics, Quantity: 5kg, Packaging: Plastic drum, CAS: 7758-11-4, White, Grade: Analysis, Melting Point: 340.0 deg.C, Molecular Formula: HK2O4P, Molecular Weight: 174.18, Percent Purity: 99+%, pH: 8.5 to 9.6 (5% soln. at 20 deg.C)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.18
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340.0°C
Color White
pH 8.5 to 9.6 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 5 kg
Assay Percent Range 99+%
Linear Formula K2HPO4
Merck Index 15,7779
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 174.18
Percent Purity ≥99%
Grade Analytical
Loss on Drying 1% (130°C) max.
Packaging Plastic drum
Chemical Name or Material Potassium phosphate, dibasic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468754 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number215472500,215470010,215470050
Lot NumberA0466047 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number215472500,215470010,215470050
Lot NumberA0399063 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number215472500,215470010,215470050
Lot NumberA0450732 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number215472500,215470010,215470050
Lot NumberA0395333 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number215472500,215470010,215470050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.