missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous Product Code.: 13256319

Potassium phosphate, dibasic, 99%, for HPLC, anhydrous

Product Code. 13256319
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 13256319

Brand: Thermo Scientific Acros 447362500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 340.0°C
CAS 7758-11-4
Quantity 250 g
Assay Percent Range 99%
Molecular Formula HK2O4P
Linear Formula K2HPO4
Merck Index 15,7779
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
Solubility Information Clear colorless
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;hydrogen phosphate
Molecular Weight (g/mol) 174.18
PubChem CID 24450
ChEBI CHEBI:32031
Formula Weight 174.18
Percent Purity 99%
Grade HPLC
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, dibasic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466787 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number447362500
Lot NumberA0463880 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number447362500
Lot NumberA0457641 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number447362500
Lot NumberA0452700 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number447362500
Lot NumberA0397481 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number447362500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.