missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™

Product Code. 10468153
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle

Potassium Phosphate Dibasic (Fine White Crystalline Powder)

This potassium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340°C
Color White
pH 8.5 to 9.6
Physical Form Solid
Quantity 1 kg
Assay Percent Range ≥98.0 %
Merck Index 15, 7779
Formula Weight 174.18
Percent Purity ≥98.0%
Identification Pass Test
Loss on Drying 1.0% max.
Packaging Poly Bottle
Decomposition Information >180°C
Chemical Name or Material Potassium Phosphate Dibasic
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'