missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Dibasic, For HPLC, ≥99.0%, Honeywell™
Click to view available options
Quantity:
250 g
Specifications
Specifications
CAS | 7758-11-4 |
Quantity | 250 g |
Molecular Formula | HK2O4P |
Linear Formula | K2HPO4 |
MDL Number | MFCD00011383 |
UN Number | NONH for all modes of transport |
Synonym | dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate |
Solubility Information | Soluble in water 0.1g/mL at 25°C |
InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
SMILES | OP(=O)([O-])[O-].[K+].[K+] |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'