missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Potassium phosphate, monobasic, 99+%, for biochemistry
£56.48 - £193.97
Chemical Identifiers
CAS | 7778-77-0 |
---|---|
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
SMILES | [K+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12694957
|
Thermo Scientific Chemicals
212595000 |
500 g |
£56.48
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12661276
|
Thermo Scientific Chemicals
212590025 |
2.5 kg |
£193.97
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
[K+].OP(O)([O-])=O |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
CHEBI:63036 |
Specifications
253.0°C | |
99+% | |
KH2PO4 | |
15,778 | |
(10% in water) Clear colorless | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
≥99% | |
Plastic bottle |
7778-77-0 | |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 | |
Biochemical | |
Potassium phosphate, monobasic |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 231-913-4
RUO – Research Use Only