missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, monobasic, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10573181

Potassium phosphate, monobasic, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10573181
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 10573181

Brand: Thermo Scientific Acros 205925000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Color Colorless to White
pH 4.2 to 4.7 (1% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula KH2PO4
Merck Index 15, 7780
Solubility Information Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 136.09
Percent Purity ≥99%
Grade Extra Pure
Loss on Drying 0.4% max. (105°C)
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, monobasic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, monobasic, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.