missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, monobasic, 99+%, pure, Thermo Scientific Chemicals
£86.60 - £240.00
Chemical Identifiers
CAS | 7778-77-0 |
---|---|
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
SMILES | [K+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10781521
|
Thermo Scientific Acros
271080025 |
2.5 kg | Plastic Bottle |
£86.60
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10185203
|
Thermo Scientific Acros
271080100 |
10 kg | Plastic drum |
£240.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
[K+].OP(O)([O-])=O |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
CHEBI:63036 |
Specifications
253.0°C | |
White | |
99+% | |
KH2PO4 | |
15, 7780 | |
Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
≥99% | |
0.5% max. (130°C) |
7778-77-0 | |
Crystalline Powder and/or Crystals | |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 | |
Pure | |
Potassium phosphate, monobasic |
Safety and Handling
missing translation for 'einecsNumber' : 231-913-4
RUO – Research Use Only