missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Monobasic, Crystal, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15508104
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Crystals
Quantity 500 g
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Grade Multi-Compendial/NF
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Chemical Name or Material Potassium Phosphate, Monobasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'