missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™

Artikelnummer. 15630730
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Meets analytical specification of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Spezifikation

Quantity 6 x 500 g
Linear Formula KH2PO4
UN Number NONH for all modes of transport
Solubility Information 4.5parts soluble in water, insoluble in alcohol
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Percent Purity 98 to 100.5% (calc. to the dried substance)
Density 2.338g/mL at 25°C
Chemical Name or Material Potassium Phosphate Monobasic, Meets analytical specification of Ph. Eur.NF
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'