missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate monobasic, meets analytical spec. of NF, Ph. Eur., E340, Anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 4 x 5 kg |
Linear Formula | KH2PO4 |
UN Number | NONH for all modes of transport |
Solubility Information | 4.5parts soluble in water, insoluble in alcohol |
IUPAC Name | potassium;dihydrogen phosphate |
Formula Weight | 136.09 |
Percent Purity | 98 to 100.5% (calc. to the dried substance) |
Density | 2.338g/mL at 25°C |
Chemical Name or Material | Potassium Phosphate Monobasic, Meets analytical specification of Ph. Eur.NF |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'