missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monobasic, for HPLC, Honeywell Fluka™

for HPLC,≥99.5%

£62.82 - £892.24

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15665940
View Documents
Honeywell-Fluka
60221-50G
50 g
£62.82
50g
Please to purchase this item. Need a web account? Register with us today!
15655940
View Documents Promotion Details
Honeywell-Fluka
60221-250G
250 g
£221.68
250g
Please to purchase this item. Need a web account? Register with us today!
15645940
View Documents
Honeywell-Fluka
60221-1KG
1 kg
£892.24
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium;dihydrogen phosphate
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
CHEBI:63036
[K+].OP(O)([O-])=O

Specifications

7778-77-0
KH2PO4
NONH for all modes of transport
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
136.09g/mol
2.338g/mL at 25°C
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
≥99.5%
Potassium phosphate monobasic
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : TC6615500

SDS
Documents

Documents

Product Certifications