missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate NF GenAR Crystal, Macron Fine Chemicals™

Product Code. 15588434
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Physical Form Crystalline
Quantity 2.5 kg
Formula Weight 136.09
Grade NF
Packaging Poly
Chemical Name or Material Potassium Phosphate, Monobasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'