missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals Product Code.: 10349962

Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10349962
1 kg, Plastic bottle
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

Product Code. 10349962

Brand: Thermo Scientific Acros 387680010

Please to purchase this item. Need a web account? Register with us today!

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.27
MDL Number MFCD00036295
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
Synonym potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
PubChem CID 62657
IUPAC Name tripotassium;phosphate
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340.0°C
Color White
Physical Form Powder, Granules or Chunks
Quantity 1 kg
Assay Percent Range 97%
Linear Formula K3PO4
Merck Index 15,7781
Solubility Information Solubility in water: 508g/L (25°C). Other solubilities: insoluble in alcohol
Formula Weight 212.27
Percent Purity 97%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, tribasic, Tribasic 97%, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0416726 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number
Lot NumberA0425039 Certificate TypeCertificate of Analysis Date17/08/2023 Catalog Number387685000,387680100,387680025,387680250,387680010
Lot NumberA0410826 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0406375 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0438936 Certificate TypeCertificate of Analysis Date09/12/2022 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.