missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, tribasic monohydrate, 96%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1340.0°C |
Color | White |
Physical Form | Powder |
Quantity | 5 kg |
Assay Percent Range | 96% |
Linear Formula | K3PO4·H2O |
Solubility Information | Solubility in water: soluble. Other solubilities: insoluble in alcohol |
Formula Weight | 230.28 |
Percent Purity | 96% |
Grade | Extra Pure |
Show More |
Safety and Handling
Product Identifier
- Potassium phosphate tribasic monohydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- H318-Causes serious eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction