missing translation for 'onlineSavingsMsg'
Learn More
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals Product Code.: 11334789

Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals

Product Code. 11334789
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11334789

Brand: Thermo Scientific Alfa Aesar 068107.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium selenocyanate is used to prepare triphenylphosphine selenide by reacting with triphenylphosphine. It finds application in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury thiocyanate and selenocyanate compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3425-46-5
Molecular Formula CKNSe
Molecular Weight (g/mol) 144.087
MDL Number MFCD00011366
InChI Key KYEKHFSRAXRJBR-UHFFFAOYSA-M
Synonym potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g
PubChem CID 76960
IUPAC Name potassium;selenocyanate
SMILES C(#N)[Se-].[K+]

Specifications

Melting Point 162°C to 164°C
Color White
Physical Form Crystalline
Quantity 10 g
UN Number UN3283
Solubility Information Soluble in water,ethanol,dimethyl formamide,hexamethylphosphoramide,acetonitrile and methanol. Slightly soluble in terahydrofuran.
Formula Weight 144.08
Percent Purity 98.5%
Odor Odorless
Sensitivity Moisture sensitive
Density 2.347 g/mL
Chemical Name or Material Potassium selenocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.