missing translation for 'onlineSavingsMsg'
Learn More
Potassium sodium tartrate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10721861

Potassium sodium tartrate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10721861
1 kg, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10721861

Brand: Thermo Scientific Acros 202860010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-59-5
MDL Number MFCD00150989
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Decomposition Information >100°C
Color White
pH 5.5 to 8.5 (5% soln. at 20°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C4H12KNaO10
Linear Formula KO2CCH(OH)CH(OH)CO2Na·4H2O
Quantity 1 kg
Beilstein 03, 525
Merck Index 15, 7790
Specific Rotation Condition + 22.10 (20.00°C c=10,water)
Molecular Weight (g/mol) 282.218
Formula Weight 282.22
Percent Purity 99 to 102%
Grade Analysis
Physical Form Crystals
Chemical Name or Material Potassium sodium tartrate tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464759 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number202860051,202860250,202865000,202860010
Lot NumberB0153967 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number202860051,202860250,202865000,202860010
Lot NumberA0462491 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number202860051,202860250,202865000,202860010
Lot NumberA0462087 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number202860051,202860250,202865000,202860010
Lot NumberA0461556 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number202860051,202860250,202865000,202860010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium sodium tartrate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.