Learn More
Potassium, solid, 99% (metals basis), Thermo Scientific Chemicals
£157.79 - £231.15
Chemical Identifiers
CAS | 7440-09-7 |
---|---|
Molecular Formula | K |
Molecular Weight (g/mol) | 39.10 |
MDL Number | MFCD00133776 |
InChI Key | ZLMJMSJWJFRBEC-UHFFFAOYSA-N |
Synonym | kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom |
PubChem CID | 5462222 |
ChEBI | CHEBI:26216 |
IUPAC Name | potassium |
SMILES | [K] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15412467
|
Thermo Scientific Alfa Aesar
045842.03 |
1 g |
£157.79
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422467
|
Thermo Scientific Alfa Aesar
045842.06 |
5 g |
£231.15
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag. This chemical is important in neuron (brain and nerve) function, and in influencing osmotic balance between cells and the interstitial fluid, with their distribution mediated in all animals (but not in all plants) by the so-called Na+/K+-ATPase pump. It is also used to prevent muscle contraction and the sending of all nerve impulses in animals through action potentials. Potassium ion is a nutrient necessary for human life and health. It is used as reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-09-7 | |
39.10 | |
ZLMJMSJWJFRBEC-UHFFFAOYSA-N | |
5462222 | |
potassium |
K | |
MFCD00133776 | |
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom | |
CHEBI:26216 | |
[K] |
Specifications
7440-09-7 | |
MFCD00133776 | |
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom | |
ZLMJMSJWJFRBEC-UHFFFAOYSA-N | |
potassium | |
5462222 | |
99.95% | |
Breakseal Ampules | |
Potassium |
K | |
UN2257 | |
Soluble in water. | |
[K] | |
39.10 | |
CHEBI:26216 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: POTASSIUM
missing translation for 'einecsNumber' : 231-119-8
missing translation for 'rtecsNumber' : TS6460000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only