missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£64.65 - £104.00
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12942185
|
Fisher Chemical
A 039 100 |
100 mL | Bottle |
£64.65
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12952185
|
Fisher Chemical
A 039 250 |
250 mL | Bottle |
£72.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12962185
|
Fisher Chemical
A 039 500 |
500 mL | Bottle |
£104.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
nitric acid |
Specifications
1g/L | |
Single-component Standard Solution | |
MFCD00011349 | |
O[N+]([O-])=O | |
63.01 | |
Liquid |
7697-37-2 | |
HNO3 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
H2O | |
Potassium |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)