Learn More
Chemical Identifiers
CAS | 24634-61-5 |
---|---|
Molecular Formula | C6H7KO2 |
Molecular Weight (g/mol) | 150.22 |
InChI Key | CHHHXKFHOYLYRE-STWYSWDKSA-M |
Synonym | potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c |
PubChem CID | 23676745 |
ChEBI | CHEBI:77868 |
IUPAC Name | potassium;(2E,4E)-hexa-2,4-dienoate |
SMILES | CC=CC=CC(=O)[O-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10126923
|
Thermo Scientific Acros
291741000 |
100 g | Plastic bottle |
£22.91
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10452825
|
Thermo Scientific Acros
291742500 |
250 g | Plastic bottle |
£38.09
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10541321
|
Thermo Scientific Acros
291740010 |
1 kg | Plastic bottle |
£88.24
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984961
|
Thermo Scientific Acros
291740050 |
5 kg | Plastic drum |
£291.45
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24634-61-5 | |
150.22 | |
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c | |
CHEBI:77868 | |
CC=CC=CC(=O)[O-].[K+] |
C6H7KO2 | |
CHHHXKFHOYLYRE-STWYSWDKSA-M | |
23676745 | |
potassium;(2E,4E)-hexa-2,4-dienoate |
Specifications
Passes test | |
270.0°C | |
1% (105°C, 3 hrs) max. | |
99% | |
CH3CH=CHCH=CHCOOK | |
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c | |
CHHHXKFHOYLYRE-STWYSWDKSA-M | |
potassium;(2E,4E)-hexa-2,4-dienoate | |
23676745 | |
150.22 | |
Powder |
24634-61-5 | |
White to Cream | |
Authentic | |
C6H7KO2 | |
15, 8847 | |
Solubility in water: 58.2 % (20°C). Other solubilities: soluble in alcohols: 6.5% (20°C) | |
CC=CC=CC(=O)[O-].[K+] | |
150.22 | |
CHEBI:77868 | |
99% | |
Potassium sorbate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 246-376-1
- Potassium sorbate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.