missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, 99+%, ACS reagent
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifikationer
Specifikationer
| Boiling Point | 1689.0°C |
| Melting Point | 1067.0°C |
| Color | White |
| pH | 5.5 to 8.5 (5% soln. at 25°C) |
| Physical Form | Fine Crystals or Powder |
| Quantity | 5 kg |
| Assay Percent Range | 99+% |
| Linear Formula | K2SO4 |
| Merck Index | 15, 7793 |
| Solubility Information | Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol |
| Visa mer |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering