missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, Certified AR for Analysis, Fisher Chemical™

£80.55 - £469.00

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10214590
View Documents Promotion Details
Fisher Chemical
P/7000/53
500 g
£80.55
500g
Please to purchase this item. Need a web account? Register with us today!
10294630
View Documents Promotion Details
Fisher Chemical
P/7000/60
1 kg
£121.00
1kg
Please to purchase this item. Need a web account? Register with us today!
10509580
View Documents
Fisher Chemical
P/7000/65
5 kg
£469.00
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium Sulfate, Certified AR for Analysis

Specifications

Chemical Identifiers

7778-80-5
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
[K+].[K+].[O-]S([O-])(=O)=O
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
CHEBI:32036

Specifications

1689°C
7778-80-5
6 to 8
K2O4S
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
[K+].[K+].[O-]S([O-])(=O)=O
174.25
CHEBI:32036
Plastic powder jar
1067°C
White
Solid
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
24507
174.25g/mol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.