missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, Honeywell™
Reagent Grade,≥99.0%
£15.61 - £42.58
Chemical Identifiers
CAS | 7778-80-5 |
---|---|
Molecular Formula | K2O4S |
Molecular Weight (g/mol) | 174.25 |
MDL Number | MFCD00011388 |
InChI Key | OTYBMLCTZGSZBG-UHFFFAOYSA-L |
Synonym | potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt |
PubChem CID | 24507 |
ChEBI | CHEBI:32036 |
IUPAC Name | dipotassium sulfate |
SMILES | [K+].[K+].[O-]S([O-])(=O)=O |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7778-80-5 | |
174.25 | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
24507 | |
dipotassium sulfate |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
CHEBI:32036 | |
[K+].[K+].[O-]S([O-])(=O)=O |
Specifications
7778-80-5 | |
K2SO4 | |
NONH for all modes of transport | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
dipotassium sulfate | |
24507 | |
174.26g/mol | |
Potassium sulfate |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
[K+].[K+].[O-]S([O-])(=O)=O | |
174.25 | |
CHEBI:32036 | |
≥99% |
Safety and Handling
missing translation for 'rtecsNumber' : TT5900000